Learn More
ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 240 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
9395.00 SEK - 11280.00 SEK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Sterility | Non-sterile |
No. per Pack | 1 Ea. |
Thickness (Metric) | 15.0 ± 0.5 mm |
Diameter (Metric) Membrane | 240 mm |
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
16863256
|
ROBU VitraPOR™
62400 |
0 |
9395.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16873256
|
ROBU VitraPOR™
62401 |
1 |
9395.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16883256
|
ROBU VitraPOR™
62402 |
2 |
9395.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16893256
|
ROBU VitraPOR™
62403 |
3 |
9395.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16803266
|
ROBU VitraPOR™
62404 |
4 |
11280.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Highly resistant to most chemicals and temperatures up to 500°C
- The biplane version may be required in specific installation circumstances
Specifications
Borosilicate Glass (3.3) | |
1 Ea. | |
240 mm |
Non-sterile | |
15.0 ± 0.5 mm |