Learn More
ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 80 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
Brand:  ROBU VitraPOR™ 16809
Additional Details : Weight : 0.24220kg
 View more versions of this product
Product Code. 16882876
  • 2450.00 SEK / Pack of 5
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications
Specifications
Borosilicate Glass (3.3)
Non-sterile
5.0 ± 0.2 mm
00
00
80 mm