Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 240 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand:  ROBU VitraPOR™ 52404
Additional Details : Weight : 1.30620kg
 View more versions of this product
Product Code. 16883156
  • 8040.00 SEK / Each
Estimated Shipment: 14-04-2023
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass 3.3
1 Ea.
19.0 ± 4.0 mm
Non-sterile
4
240 mm