Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 270 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand: ROBU VitraPOR™ 52701
Additional Details : Weight : 1.83350kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Non-sterile | |
1 | |
270 mm |
1 μm | |
1 Ea. | |
26.0 ± 6.0 mm |