missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 350 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 53502

Additional Details : Weight : 3.61000kg

 View more versions of this product

Product Code. 16813196

  • 16090.00 SEK / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
1 Ea.
26.0 ± 6.0 mm
Non-sterile
2
350 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers