Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 350 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand: ROBU VitraPOR™ 53504
Additional Details : Weight : 3.61000kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
1 Ea. | |
26.0 ± 6.0 mm |
Non-sterile | |
4 | |
350 mm |