Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 5 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand: ROBU VitraPOR™ 15051
Additional Details : Weight : 0.00480kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Non-sterile | |
1 | |
5 mm |
1 μm | |
10/Pk. | |
2.8 ± 0.7 mm |