Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 5 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
472.00 SEK - 1565.00 SEK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Sterility | Non-sterile |
No. per Pack | 10/Pk. |
Thickness (Metric) | 2.8 ± 0.7 mm |
Diameter (Metric) Membrane | 5 mm |
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
16862756
|
ROBU VitraPOR™
15052 |
2 |
472.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16882756
|
ROBU VitraPOR™
15054 |
4 |
540.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16872756
|
ROBU VitraPOR™
15053 |
3 |
472.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16892756
|
ROBU VitraPOR™
15055 |
5 |
1565.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16852756
|
ROBU VitraPOR™
15051 |
1 |
472.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16842756
|
ROBU VitraPOR™
15050 |
0 |
472.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16802766
|
ROBU VitraPOR™
1505F |
Fine |
635.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
10/Pk. | |
5 mm |
Non-sterile | |
2.8 ± 0.7 mm |