Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 5 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand:  ROBU VitraPOR™ 15055
Additional Details : Weight : 0.00480kg
 View more versions of this product
Product Code. 16892756
  • 1565.00 SEK / Pack of 10
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
10/Pk.
2.8 ± 0.7 mm
Non-sterile
5
5 mm