Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 80 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand: ROBU VitraPOR™ 15802
Additional Details : Weight : 0.24220kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
5/Pk. | |
6.0 ± 1.4 mm |
Non-sterile | |
2 | |
80 mm |