Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 80 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand:  ROBU VitraPOR™ 15805
Additional Details : Weight : 0.24220kg
 View more versions of this product
Product Code. 16882796
  • 4945.00 SEK / Pack of 5
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
5/Pk.
6.0 ± 1.4 mm
Non-sterile
5
80 mm