Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 80 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand:  ROBU VitraPOR™ 15809
Additional Details : Weight : 0.24220kg
 View more versions of this product
Product Code. 16892796
  • 1515.00 SEK / Pack of 5
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
Non-sterile
00
80 mm
00
5/Pk.
6.0 ± 1.4 mm