missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Filter Funnel with Standard Joint
Filter Funnel with fused filter disc.
Brand: ROBU VitraPOR™ 47023
This item is not returnable.
View return policy
Description
- With fused filter disc
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
5 mL | |
39 mm | |
3 |
Borosilicate Glass (3.3) | |
NSK 14/23 | |
2/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ROBU VitraPOR™ Filter Funnel with Standard Joint > NSK 14/23; Pore Size: 3
Spot an opportunity for improvement?Share a Content Correction