Learn More
Fisherbrand™ Combs for Mini-Plus Wide Format Horizontal Gel Units, 1.5mm Thick
For use with Mini-Plus Wide Format Horizontal Gel Units.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11513372
  • 391.00 SEK / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Mini-Plus Wide Format Horizontal Gel Units
8
1.5 mm
Comb
1 Each
100 μL