Learn More
Fisherbrand™ Polypropylene, Rectangular Carboy
For Use With: Ideal for storing and dispensing lab water and reagents; Autoclavable; Closure Material: Polypropylene
Brand:  Fisherbrand™ 302212-0020
Product Code. 13448029
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Value your lab can rely on. Fisherbrand Polypropylene Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
Ideal for storing and dispensing lab water and reagents
  • Durable construction for long service life
  • Molded graduations for 1 gallon and 51 increments
  • Choice of materials for general use or autoclavability
  • Space-saving rectangular design
  • Wide mouth closure allows for easy filling and cleaning access
  • Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
  • Spigot spout without the adapter fits 5/8-in. ID tubing
  • Materials meet FDA requirements for food use and USP VI criteria (non-toxic)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
9 L
Polypropylene
Ideal for storing and dispensing lab water and reagents
Wide
Rectangular
2.38 gal.
FDA, USP VI
White
Polypropylene
Carboy
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Polypropylene, Rectangular Carboy > 2.38 gal. (9L); Polypropylene; Without spigot