missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Five elements solution, Ca, Fe, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical MS009050
This item is not returnable.
View return policy
Specifications
Five Elements | |
Ca,Fe,Mg,K,Na | |
1 g/L±0.002 g/L | |
Ca, Fe, Mg, K, Na | |
50 mL |
Bottle | |
Liquid | |
ICP/MS Calibration Multi-element Standard Solution | |
2% HNO3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Five elements solution, Ca, Fe, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™ >
Spot an opportunity for improvement?Share a Content Correction