Learn More
Five elements solution, Ca, Fe, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand:  Fisher Chemical MS 009 100
 View more versions of this product
Product Code. 12976193
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Five Elements
Ca,Fe,Mg,K,Na
1 g/L±0.002 g/L
Ca, Fe, Mg, K, Na
100 mL
Bottle
Liquid
ICP/MS Calibration Multi-element Standard Solution
2% HNO3
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Five elements solution, Ca, Fe, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™ >