Learn More
Five elements solution, Ca, Fe, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand:  Fisher Chemical MS 009 250
 View more versions of this product
Product Code. 12986193
Specifications
Five Elements
Ca,Fe,Mg,K,Na
1 g/L±0.002 g/L
Ca, Fe, Mg, K, Na
250 mL
Bottle
Liquid
ICP/MS Calibration Multi-element Standard Solution
2% HNO3
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Five elements solution, Ca, Fe, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™ >