missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FKBP1A (Human) Recombinant Protein
Human FKBP1A full-length ORF ( AAH05147, 1 a.a. - 108 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002280-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGVQVETISPGDGRTFPKRGQTCVVHYTGMLEDGKKFDSSRDRNKPFKFMLGKQEVIRGWEEGVAQMSVGQRAKLTISPDYAYGATGHPGIIPPHATLVFDVELLKLE
Specifications
AAH05147 | |
FK506 binding protein 1A, 12kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKBP1A | |
GST |
2280 | |
Wheat germ expression system | |
25 μg | |
FKBP-12, FKBP1, FKBP12, FKBP12C, PKC12, PKCI2, PPIASE | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |