missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FKBP1A (Human) Recombinant Protein
Human FKBP1A full-length ORF ( AAH05147, 1 a.a. - 108 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH05147 |
---|---|
Gene ID (Entrez) | 2280 |
Name | FK506 binding protein 1A, 12kDa |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135011
|
Abnova™
H00002280-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16145011
|
Abnova™
H00002280-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MGVQVETISPGDGRTFPKRGQTCVVHYTGMLEDGKKFDSSRDRNKPFKFMLGKQEVIRGWEEGVAQMSVGQRAKLTISPDYAYGATGHPGIIPPHATLVFDVELLKLE
Specifications
AAH05147 | |
FK506 binding protein 1A, 12kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
FKBP-12, FKBP1, FKBP12, FKBP12C, PKC12, PKCI2, PPIASE | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2280 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKBP1A | |
GST |