Learn More
Flamingo homolog 1/CELSR3 Mouse anti-Human, Alexa Fluor 594, Clone: 763103, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7278T-100UG

Description
Flamingo homolog 1/CELSR3 Monoclonal specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Flow Cytometry.Specifications
Flamingo homolog 1/CELSR3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9NYQ7 | |
CELSR3 | |
E. coli-derived recombinant human CELSR3 Val531-Ser711 Accession # Q9NYQ7 | |
RUO | |
1951 | |
Human | |
IgG2b |
Flow Cytometry | |
763103 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11, CDHF11RESDA1, EGFL1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2, RESDA1 | |
Mouse | |
100 ÎĽg | |
Primary | |
Detects human CELSR3 in ELISA. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only (RUO)