Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Narrow Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07045070
 View more versions of this product
Product Code. 10080401
  • 3725.00 SEK / Pack of 10
Only null left
Estimated Shipment: 25-07-2025
Description
  • Acc. to DIN ISO 1773 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Specifications
1 L
Duran Glass
190 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente Flat Bottom Flasks, Narrow Neck > 1000 mL