Learn More
Lenz Laborglasintrumente Flat-Bottom Flasks With Conical Ground Joint
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03012437
 View more versions of this product
Product Code. 10500122
  • 1515.00 SEK / Pack of 10
Only null left
Estimated Shipment: 25-07-2025
Description
  • According to DIN EN ISO 4797
  • Wiith inscription label
  • Material: DURAN™
Specifications
Specifications
100 mL
NS 24/29
10/Pk.
Conical ground joint and inscription label
64 mm
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente Flat-Bottom Flasks With Conical Ground Joint > 100 mL