missing translation for 'onlineSavingsMsg'
Learn More

FLI1 Mouse, Clone: FLI1/1312, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 2313-MSM2-P0

 View more versions of this product

Product Code. 15949997

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

FLI1
Nuclear
FLI1/1312
Unconjugated
NP_001161153
Mouse
51kDa
Protein A/G Purified
RUO
2313
IgG
Western Blot
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
FLI1
Recombinant fragment (aa 107-261) of human FLI1 protein
THP1 and Raji cells. Tonsil, Ewing s sarcoma (EWS) and Angiosarcoma
20 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FLI1 Mouse, Clone: FLI1/1312, enQuireBio™ > 20 μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.