Learn More
Abnova™ FLI1 Split FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Specifications
Specifications
Content And Storage | Store at 4°C in the dark |
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | Friend leukemia virus integration 1 |
Gene Alias | EWSR2,SIC-1 |
Gene Symbol | FLI1 |
Gene ID (Entrez) | 2313 |
Product Type | DNA/RNA |
Quantity | 100 μL |
Form | Liquid |
Product Content Correction
Product Title
Abnova™ FLI1 Split FISH Probe >
Spot an opportunity for improvement?Share a Content Correction