Learn More
FLII Mouse anti-Human, Unconjugated, Clone: 2F9C8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67039-1-IG-150UL
 View more versions of this product
Product Code. 16879425
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The FLII antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FLII. This antibody recognizes human, mouse, rat antigen. The FLII antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
FLII
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
2F9C8
Unconjugated
150 μL
2314
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FLII Mouse anti-Human, Unconjugated, Clone: 2F9C8, Proteintech > 150 μL