Learn More
FLII Mouse anti-Human, Unconjugated, Clone: 2F9C8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67039-1-IG-150UL
 View more versions of this product
Product Code. 16879425
  • 4652.90 SEK / 150µL
Description
The FLII antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FLII. This antibody recognizes human, mouse, rat antigen. The FLII antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
FLII
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
2F9C8
Unconjugated
150 μL
2314
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
FLII Mouse anti-Human, Unconjugated, Clone: 2F9C8, Proteintech > 150 μL