missing translation for 'onlineSavingsMsg'
Learn More
Fluorolink∣r E10-H Product Code.: 15412739

Fluorolink∣r E10-H

Product Code. 15412739
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15412739

Brand: Thermo Scientific Alfa Aesar H66374.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use in polymer modifications | CAS: 162492-15-1

Use in polymer modifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use in polymer modifications.

Solubility
Soluble in fluorinated solvents, alcohol, and ketones. Insoluble in water.

Notes
Incompatible with heat and oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162492-15-1

Specifications

Color Colorless to Yellow
Density 1.73 g/mL
Odor Odorless
Refractive Index 1.317
Quantity 25 g
Solubility Information Soluble in fluorinated solvents,alcohol,and ketones; Insoluble in water.
Physical Form Liquid
Chemical Name or Material Fluorolink™ E10-H

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberY01K023 Certificate TypeCertificate of Analysis Date11/02/2025 Product CodeH66374.22,H66374.14
Lot NumberW22J006 Certificate TypeCertificate of Analysis Date06/10/2023 Product CodeH66374.22,H66374.14
Lot NumberS16I031 Certificate TypeCertificate of Analysis Date25/07/2022 Product CodeH66374.22,H66374.14
Lot NumberS30I018 Certificate TypeCertificate of Analysis Date20/07/2022 Product CodeH66374.22,H66374.14
Lot NumberZ02H049 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeH66374.22,H66374.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fluorolink∣r E10-H >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.