Learn More
FOXA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20411-1-AP-20UL
 View more versions of this product
Product Code. 16879944
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
Specifications
Specifications
FOXA1
Polyclonal
Unconjugated
FOXA1
forkhead box A1, Forkhead box protein A1, FOXA1, HNF 3 alpha, HNF 3A, HNF3A, TCF 3A, TCF3A, Transcription factor 3A
Rabbit
Antigen Affinity Chromatography
RUO
15375, 3169
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35582, P55317
FOXA1
FOXA1 Fusion Protein Ag14243
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FOXA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated