missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1395.65 SEK - 3774.95 SEK
Specifications
Antigen | FOXB1 |
---|---|
Concentration | 0.51 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Foxb1 gene ontology annotations related to this gene include anatomical structure morphogenesis; axon target recognition; cell differentiation; cell migration in diencephalon; epithelial cell differentiation involved in mammary gland alveolus development; floor plate development; hypothalamus cell migration; inferior colliculus development; lactation; mammary gland lobule development; mammillary body development; mammillothalamic axonal tract development; midbrain development; negative regulation of neuron apoptotic process; regulation of transcription by RNA polymerase II; somitogenesis; spinal cord development; telencephalon cell migration; thalamus development; urogenital system development; visual learning.Specifications
FOXB1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q64732, Q99853 | |
27023, 64290 | |
FOXB1 Fusion Protein Ag18079 | |
Primary | |
-20°C | |
FOXB1 |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FKH5, forkhead box B1, Forkhead box protein B1, FOXB1, HFKH 5, Transcription factor FKH 5 | |
Foxb1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FOXB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech