missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXD4L6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | FOXD4L6 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881625
|
Proteintech
22081-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871625
|
Proteintech
22081-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5 FOXD4-like proteins (FOXD4L2, FOXD4L3, FOXD4L4, FOXD4L5 and FOXD4L6) are encoded by a strongly repeated region of the 9q12 chromosome region. They are very similar and it is therefore difficult to provide a clear and unambiguous protein sequence. Our sequences are in agreement with HGNC nomenclature.Specifications
FOXD4L6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q3SYB3 | |
653404 | |
FOXD4L6 Fusion Protein Ag17247 | |
Primary | |
-20°C | |
FOXD4L6 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
forkhead box D4 like 6, Forkhead box protein D4 like 6, FOXD4 like 6, FOXD4L6 | |
FOXD4L6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FOXD4L6 Rabbit anti-Human, Polyclonal, Proteintech