Learn More
Abnova™ FOXP1 Split FISH Probe Product Code.: 16033475
Abnova™ FOXP1 Split FISH Probe
Product Code. 16033475
200 μL, 200µL
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Product Code. 16033475
Brand: Abnova™ FS0041.200uL
Includes
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Specifications
Content And Storage Store at 4°C in the dark.
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Format Tube
For Use With (Application) FISH-Ce
Gene ID (Entrez) 27086
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Quantity 200 μL
Form Liquid
Product Content Correction
Product Title
Abnova™ FOXP1 Split FISH Probe >