missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16234-1-AP-150UL
This item is not returnable.
View return policy
Description
The forkhead family of transcription factors belongs to the winged helix class of DNA-binding proteins. The protein encoded by this intronless gene contains a forkhead domain and is found predomitly in aorta and kidney. The function of the encoded protein is unknown.Specifications
FOXS1 | |
Polyclonal | |
Unconjugated | |
FOXS1 | |
FKHL18, Forkhead box protein S1, forkhead box S1, Forkhead like 18 protein, FOXS1, FREAC 10, FREAC10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14239, 2307, 311547 | |
-20°C | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43638, Q61574 | |
Foxs1 | |
FOXS1 Fusion Protein Ag8957 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FOXS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction