missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled 9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13865-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the ′frizzled′ gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predomitly in brain, testis, eye, skeletal muscle, and kidney.Specifications
Frizzled 9 | |
Polyclonal | |
Unconjugated | |
FZD9 | |
CD349, Frizzled 9, Fz 9, FZD3, FZD9, FzE6, hFz9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14371, 266608, 8326 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00144, Q8K4C8, Q9R216 | |
Fzd9 | |
Frizzled 9 Fusion Protein Ag4847 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Frizzled 9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction