missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSTL1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20182-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein with similarity to follistatin, an activin-binding protein. It contains an FS module, a follistatin-like sequence containing 10 conserved cysteine residues. This gene product is thought to be an autoantigen associated with rheumatoid arthritis.Specifications
FSTL1 | |
Polyclonal | |
Unconjugated | |
FSTL1 | |
follistatin like 1, Follistatin like protein 1, Follistatin related protein 1, FRP, FSL1, FSTL1, TSC-36 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11167, 79210 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12841, Q62632 | |
Fstl1 | |
FSTL1 Fusion Protein Ag14057 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |