Learn More
FUCA1 Mouse anti-Human, Unconjugated, Clone: 1G5C5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67251-1-IG-20UL
 View more versions of this product
Product Code. 16889775
  • 1645.54 SEK / 20µL
Description
The FUCA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FUCA1. This antibody recognizes human, mouse antigen. The FUCA1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
FUCA1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1G5C5
Unconjugated
20 μL
2517
IgG2b
Certificates
Promotions
Promotions
Product Content Correction
Product Title
FUCA1 Mouse anti-Human, Unconjugated, Clone: 1G5C5, Proteintech > 20 μL