Learn More
FUCA1 Mouse anti-Human, Unconjugated, Clone: 1G5C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67251-1-IG-20UL
Description
The FUCA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FUCA1. This antibody recognizes human, mouse antigen. The FUCA1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
FUCA1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1G5C5 | |
Unconjugated | |
20 μL | |
2517 | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.