missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17956-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
FUT1 is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway.Specifications
FUT1 | |
Polyclonal | |
Unconjugated | |
FUT1 | |
Alpha(1,2)FT 1, Fucosyltransferase 1, FUT1, H, HH, HSC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14343, 2523 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O09160, P19526 | |
FUT1 | |
FUT1 Fusion Protein Ag12450 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |