missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | FUT11 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887014
|
Proteintech
17175-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877014
|
Proteintech
17175-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Probable fucosyltransferase.Specifications
FUT11 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q495W5, Q68FV3, Q8BHC9 | |
170384, 286971, 73068 | |
FUT11 Fusion Protein Ag10815 | |
Primary | |
-20°C | |
FUT11 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Fuc TXI, Fucosyltransferase 11, Fucosyltransferase XI, FucT XI, FUT11 | |
Fut11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FUT11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech