Learn More
A&D Weighing FX/FZ Precision Balances
A&D Weighing FZ/FX Series Precision Balances combine all of the essential weighing functions in a compact, light-weight package.
7672.09 SEK - 9911.25 SEK
Specifications
Certifications/Compliance | GLP/GMP/GCP/ISO |
---|---|
Display Type | VFD |
Material | Stainless Steel |
Stabilization Time | 1 second |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
16601072
|
A&D Weighing
FX-120I |
122 g |
7672.09 SEK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16601082
|
A&D Weighing
FX-5000I |
5 kg |
9840.54 SEK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16611082
|
A&D Weighing
FX-500I |
520 g |
9911.25 SEK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16621072
|
A&D Weighing
FX-2000I |
2200 g |
8267.24 SEK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16641072
|
A&D Weighing
FX-200I |
220 g |
8267.24 SEK
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16661072
|
A&D Weighing
FX-3000I |
3200 g |
8768.10 SEK
Each |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16681062
|
A&D Weighing
FX-1200I |
1220 g |
7672.09 SEK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16681072
|
A&D Weighing
FX-300I |
320 g |
8768.10 SEK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Built-in internal calibration at the touch of a button
- Shock absorber function
- Auto power ON/OFF function
- B5 size footprint for use in narrow spaces
- VFD Display for easy to read weighing results
- SCF – Statistical Calculation Function
- Standard RS-232C
- Fast compact SHS sensor, response time only 1 second
- Quick USB interface – no driver installation required (optional)
- LAN – Ethernet interface with WinCT-Plus software (optional)
- Animal weighing pan (optional)
- Large breeze break (optional)
- Small breeze break included on 1 mg models
- Built-in rechargeable NiMH battery pack (optional)
Specifications
GLP/GMP/GCP/ISO | |
VFD | |
Stainless Steel | |
1 second |