Learn More
FXYD1 Mouse anti-Human, Unconjugated, Clone: 2C10G9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67150-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16879615
  • 3665.00 SEK / 150µL
Estimated Shipment: 30-03-2023
to see stock.
Description
Description
The FXYD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FXYD1. This antibody recognizes human, pig, mouse antigen. The FXYD1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
FXYD1
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Pig, Human
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
2C10G9
Unconjugated
150 μL
5348
IgG2a