Learn More
FXYD1 Mouse anti-Human, Unconjugated, Clone: 2C10G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67150-1-IG-20UL
Description
The FXYD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FXYD1. This antibody recognizes human, pig, mouse antigen. The FXYD1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
FXYD1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Pig, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2C10G9 | |
Unconjugated | |
20 μL | |
5348 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.