missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYN Mouse anti-Human, Mouse, Rat, Clone: 1G4B1, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | FYN |
---|---|
Clone | 1G4B1 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888635
|
Proteintech
66606-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878635
|
Proteintech
66606-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.Specifications
FYN | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FYN, p59 Fyn, Proto oncogene c Fyn, Proto oncogene Syn, SLK, Src like kinase, SYN, Tyrosine protein kinase Fyn | |
Fyn | |
IgG1 | |
Protein G | |
Antibody |
1G4B1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Rat, Human, Mouse | |
P06241, P39688, Q62844 | |
14360, 25150, 2534 | |
FYN Fusion Protein Ag11127 | |
Primary | |
-20°C | |
FYN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FYN Mouse anti-Human, Mouse, Rat, Clone: 1G4B1, Proteintech