missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FZD5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21519-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the ′frizzled′ gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD5 protein is believed to be the receptor for the Wnt5A ligand.Specifications
FZD5 | |
Polyclonal | |
Unconjugated | |
FZD5 | |
C2orf31, Frizzled 5, Fz 5, FZD5, FzE5, HFZ5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7855 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13467 | |
FZD5 | |
FZD5 Fusion Protein Ag16082 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FZD5 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction