Learn More
GAB1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26200-1-AP-20UL
Description
The GAB1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human GAB1. This antibody recognizes human, rat antigen. The GAB1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
GAB1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
2549 | |
Liquid |
Western Blot, ELISA | |
600 μg/mL | |
Rabbit | |
Primary | |
Human, Rat |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.