Learn More
GAB1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26200-1-AP-20UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17231386
  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.
Description
Description
The GAB1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human GAB1. This antibody recognizes human, rat antigen. The GAB1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
GAB1
Polyclonal
Unconjugated
20 μL
2549
Liquid
Western Blot, ELISA
600 μg/mL
Rabbit
Primary
Human, Rat