Learn More
GAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19958-1-AP-150UL
 View more versions of this product
Product Code. 16889434
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This antibody is specific to GAC isoform. It has no cross reaction to KGA.
Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.
Specifications
Specifications
GAC
Polyclonal
Unconjugated
GLS
AAD20, GAC, GAC-specific, GLS, GLS1, glutaminase C, K glutaminase, KIAA0838, L glutamine amidohydrolase
Rabbit
Antigen Affinity Chromatography
RUO
14660, 24398, 2744
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3Z7P3, O94925, P13264
GLS
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated