Learn More
GAGE2D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12532-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
GAGE2D, also named as GAGE8 and CT4.8, is expressed in testis and tumor tissues.Specifications
GAGE2D | |
Polyclonal | |
Unconjugated | |
GAGE2D | |
Cancer/testis antigen 4.8, CT4.8, G antigen 2D, G antigen 8, GAGE 8, GAGE2D, GAGE8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
729408 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UEU5 | |
GAGE2D | |
GAGE2D Fusion Protein Ag3230 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |