missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAGE2D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | GAGE2D |
---|---|
Concentration | 0.57 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850284
|
Proteintech
12532-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16840284
|
Proteintech
12532-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAGE2D, also named as GAGE8 and CT4.8, is expressed in testis and tumor tissues.Specifications
GAGE2D | |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q9UEU5 | |
729408 | |
GAGE2D Fusion Protein Ag3230 | |
Primary | |
-20°C | |
GAGE2D |
0.57 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Cancer/testis antigen 4.8, CT4.8, G antigen 2D, G antigen 8, GAGE 8, GAGE2D, GAGE8 | |
GAGE2D | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GAGE2D Rabbit anti-Human, Polyclonal, Proteintech