missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAL3ST4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16240-1-AP-150UL
This item is not returnable.
View return policy
Description
Catalyzes the transfer of sulfate to beta-1,3-linked galactose residues in O-linked glycoproteins. Good substrates include asialofetuin, Gal-beta-1,3-GalNAc and Gal-beta-1,3 (GlcNAc-beta-1,6)GalNAc.Specifications
GAL3ST4 | |
Polyclonal | |
Unconjugated | |
GAL3ST4 | |
GAL3ST 4, GAL3ST4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
330217, 498166, 79690 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96RP7 | |
Gal3st4 | |
GAL3ST4 Fusion Protein Ag9063 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GAL3ST4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction