Learn More
Galectin 2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12849-2-AP-150UL
 View more versions of this product
Product Code. 16880784
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction.
Specifications
Specifications
Galectin 2
Polyclonal
Unconjugated
LGALS2
Gal 2, Galectin 2, HL14, Lactose binding lectin 2, LGALS2, S Lac lectin 2
Rabbit
Antigen Affinity Chromatography
RUO
3957
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P05162
LGALS2
Galectin 2 Fusion Protein Ag3979
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Galectin 2 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated